Please login to your online account to display your discounted pricing

4-(2-Morpholin-4-ylethoxy)aniline, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC42114DA

 View more versions of this product

Catalog No. CC42114DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 222.29
Quantity 1g
Assay 97%
Chemical Name or Material 4-(2-Morpholin-4-ylethoxy)aniline
Molecular Formula C12H18N2O2
Assay Percent Range 97%
CAS 52481-41-1
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(2-Morpholin-4-ylethoxy)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H18N2O2
Formula Weight 222.29