Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Methylphenyl)-3-thiosemicarbazide, 98%

Chemical Identifiers

Chemical Name or Material 4-(2-Methylphenyl)-3-thiosemicarbazide
CAS 614-10-8
Molecular Formula C8H11N3S
Formula Weight 181.26
MDL Number MFCD00041277
UN Number UN2811
Beilstein 880437
Synonym 4-(o-Tolyl)-3-thiosemicarbazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0938206

  SDS
alfa aesar™
L0938206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0938214

  SDS
alfa aesar™
L0938214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.26
Chemical Name or Material 4-(2-Methylphenyl)-3-thiosemicarbazide
MDL Number MFCD00041277
Assay 98%
Molecular Formula C8H11N3S
CAS 614-10-8
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 880437
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Quantity 5g
Synonym 4-(o-Tolyl)-3-thiosemicarbazide
RTECS Number VT4545000
UN Number UN2811
Melting Point 145° to 148°C
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.