Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Methylphenoxy)piperidine, 97%

$384.82 - $384.82

Chemical Identifiers

Chemical Name or Material 4-(2-Methylphenoxy)piperidine
CAS 63843-42-5
Molecular Formula C12H17NO
Formula Weight 191.28
MDL Number MFCD06248344
Synonym 4-(o-Tolyloxy)piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5095103

 
alfa aesar™
H5095103
1g Each for $384.82
Description & Specifications

Specifications

Formula Weight 191.28
Chemical Name or Material 4-(2-Methylphenoxy)piperidine
Assay 97%
Molecular Formula C12H17NO
MDL Number MFCD06248344
CAS 63843-42-5
Synonym 4-(o-Tolyloxy)piperidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.