Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide

$102.31 - $355.59

Chemical Identifiers

Chemical Name or Material 4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide
CAS 902836-97-9
Molecular Formula C14H16N2O3S
Formula Weight 292.36
MDL Number MFCD08061109
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5100303

  SDS
alfa aesar™
H5100303
1g Each for $355.59

AAH51003MD

  SDS
alfa aesar™
H51003MD
250mg Each for $102.31
Description & Specifications

Specifications

Formula Weight 292.36
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C14H16N2O3S
Health Hazard 1 Warning
CAS 902836-97-9
Quantity 1g
MDL Number MFCD08061109
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.