Please login to your online account to display your discounted pricing

4-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride, Tech., Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC40302DA

 View more versions of this product

Catalog No. CC40302DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 237.71
Quantity 1g
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzoyl chloride
Packaging Amber Glass Bottle
Molecular Formula C11H8ClNOS
CAS 857283-93-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD08271902
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

Specifications

Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzoyl chloride
CAS 857283-93-3
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C11H8ClNOS
Formula Weight 237.71
MDL Number MFCD08271902
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents