Please login to your online account to display your discounted pricing

4-(2-Methyl-1,3-thiazol-4-yl)benzenesulfonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzenesulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C10 H8 Cl N O2 S2
Formula Weight 273.76
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC40303CB

  SDS
fisher scientific company
CC40303CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.76
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10 H8 Cl N O2 S2
CAS 852180-73-5
Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzenesulfonyl chloride