Please login to your online account to display your discounted pricing

4-(2-Methyl-1,3-thiazol-4-yl)benzenesulfonyl chloride, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC40303CB

 View more versions of this product

Catalog No. CC40303CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 273.76
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10 H8 Cl N O2 S2
CAS 852180-73-5
Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzenesulfonyl chloride

Specifications

Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzenesulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C10 H8 Cl N O2 S2
Formula Weight 273.76