Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Methoxyphenyl)-1-piperazineacetamidoxime, 99%

$484.11 - $484.11

Chemical Identifiers

Chemical Name or Material 4-(2-Methoxyphenyl)-1-piperazineacetamidoxime
CAS 261623-50-1
Molecular Formula C13H20N4O2
Formula Weight 264.33g/mol
MDL Number MFCD08060956
Synonym N-Hydroxy-4-(2-methoxyphenyl)-1-piperazineacetamidine; N-Hydroxy-2-[4-(2-methoxyphenyl)piperazin-1-yl]ethanimidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5066806

 
alfa aesar™
H50668-06
5g Each for $484.11
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material 4-(2-Methoxyphenyl)-1-piperazineacetamidoxime
CAS 261623-50-1
Assay 99%
Molecular Formula C13H20N4O2
Formula Weight 264.33g/mol
MDL Number MFCD08060956
Synonym N-Hydroxy-4-(2-methoxyphenyl)-1-piperazineacetamidine; N-Hydroxy-2-[4-(2-methoxyphenyl)piperazin-1-yl]ethanimidamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.