missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Methoxyethylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53155MD

 View more versions of this product

Catalog No. AAH53155MD



Specifications

Specifications

4-(2-Methoxyethylcarbamoyl)benzeneboronic acid
250mg
C10H14BNO4
4-2-methoxyethylcarbamoyl phenylboronic acid, 4-2-methoxyethylaminocarbonyl phenylboronic acid, 4-2-methoxyethylaminocarbonyl benzeneboronic acid, 4-2-methoxyethyl carbamoyl phenylboronic acid, acmc-209q2x, 4-2-methoxyethyl carbamoyl phenyl boronic acid, 4-2-methoxyethyl aminocarbonyl benzeneboronic acid, boronic acid,b-4-2-methoxyethyl amino carbonyl phenyl, boronicacid,b-4-2-methoxyethyl amino carbonyl phenyl
B(C1=CC=C(C=C1)C(=O)NCCOC)(O)O
223.035
223.04
120°C to 124°C
850589-34-3
MFCD04115682
QRGVHOKYRIQQTN-UHFFFAOYSA-N
[4-(2-methoxyethylcarbamoyl)phenyl]boronic acid
44119550
98%

Chemical Identifiers

850589-34-3
223.035
QRGVHOKYRIQQTN-UHFFFAOYSA-N
44119550
B(C1=CC=C(C=C1)C(=O)NCCOC)(O)O
C10H14BNO4
MFCD04115682
4-2-methoxyethylcarbamoyl phenylboronic acid, 4-2-methoxyethylaminocarbonyl phenylboronic acid, 4-2-methoxyethylaminocarbonyl benzeneboronic acid, 4-2-methoxyethyl carbamoyl phenylboronic acid, acmc-209q2x, 4-2-methoxyethyl carbamoyl phenyl boronic acid, 4-2-methoxyethyl aminocarbonyl benzeneboronic acid, boronic acid,b-4-2-methoxyethyl amino carbonyl phenyl, boronicacid,b-4-2-methoxyethyl amino carbonyl phenyl
[4-(2-methoxyethylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : Y

Documents