missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Methoxyethoxy)benzeneboronic acid, 95%

$31.61 - $296.08

Chemical Identifiers

CAS 265664-52-6
Molecular Formula C9H13BO4
Molecular Weight (g/mol) 196.009
MDL Number MFCD03452784
InChI Key ODKMFCPXFMQWHA-UHFFFAOYSA-N
Synonym 4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy phenylboronic acid, 4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy benzeneboronic acid, acmc-209gsb, 4-2-methoxyethoxy phenylboronicacid, boronic acid, 4-2-methoxyethoxy phenyl
PubChem CID 23508303
IUPAC Name [4-(2-methoxyethoxy)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)OCCOC)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64050MD View Documents Alfa Aesar™
H64050MD
250mg Each for $31.61
 
AAH6405003 View Documents Alfa Aesar™
H6405003
1g Each for $79.73
 
AAH6405006 View Documents Alfa Aesar™
H6405006
5g Each for $296.08
 
Specifications

Chemical Identifiers

265664-52-6
196.009
ODKMFCPXFMQWHA-UHFFFAOYSA-N
23508303
B(C1=CC=C(C=C1)OCCOC)(O)O
C9H13BO4
MFCD03452784
4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy phenylboronic acid, 4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy benzeneboronic acid, acmc-209gsb, 4-2-methoxyethoxy phenylboronicacid, boronic acid, 4-2-methoxyethoxy phenyl
[4-(2-methoxyethoxy)phenyl]boronic acid

Specifications

250mg
C9H13BO4
4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy phenylboronic acid, 4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy benzeneboronic acid, acmc-209gsb, 4-2-methoxyethoxy phenylboronicacid, boronic acid, 4-2-methoxyethoxy phenyl
B(C1=CC=C(C=C1)OCCOC)(O)O
196.009
196.01
4-(2-Methoxyethoxy)benzeneboronic acid
265664-52-6
MFCD03452784
ODKMFCPXFMQWHA-UHFFFAOYSA-N
[4-(2-methoxyethoxy)phenyl]boronic acid
23508303
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning