Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Methoxy-4-methylphenoxy)piperidine, 97%

$265.86 - $265.86

Chemical Identifiers

Chemical Name or Material 4-(2-Methoxy-4-methylphenoxy)piperidine
CAS 883543-21-3
Molecular Formula C13H19NO2
Formula Weight 221.30g/mol
MDL Number MFCD06248038
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5096303

 
alfa aesar™
H50963-03
Each for $265.86
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-(2-Methoxy-4-methylphenoxy)piperidine
CAS 883543-21-3
Assay 97%
Molecular Formula C13H19NO2
Formula Weight 221.30g/mol
MDL Number MFCD06248038
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.