Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Hydroxyhexafluoroisopropyl)benzoic acid, 97%

$133.46 - $498.73

Chemical Identifiers

Chemical Name or Material 4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
CAS 16261-80-6
Molecular Formula C10H6F6O3
Formula Weight 288.15
MDL Number MFCD00236043
Beilstein 2148278
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1455403

  SDS
alfa aesar™
L1455403
1g Each for $133.46

AAL1455406

  SDS
alfa aesar™
L1455406
5g Each for $498.73
Description & Specifications

Specifications

Formula Weight 288.15
Chemical Name or Material 4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
CAS 16261-80-6
Health Hazard 1 Warning
Assay 97%
Molecular Formula C10H6F6O3
MDL Number MFCD00236043
Beilstein 2148278
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Odorless
Melting Point 122° to 126°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
4-(2-Hydroxyhexafluoroisopropyl) benzoic acid is used in chemicals, pharmaceutical research, acts as a pharmaceutical intermediate and reagent.

Notes
Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.