Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester, 95%

$70.68 - $286.28

Chemical Identifiers

Chemical Name or Material 4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester
CAS 1206641-45-3
Molecular Formula C16H26BNO3
Formula Weight 291.2
MDL Number MFCD16294544
Synonym 2-(Methyl[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]amino)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5194106

  SDS
alfa aesar™
H5194106
5g Each for $286.28

AAH5194103

  SDS
alfa aesar™
H5194103
1g Each for $70.68
Description & Specifications

Specifications

Chemical Name or Material 4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester
Quantity 5g
CAS 1206641-45-3
Assay 95%
Molecular Formula C16H26BNO3
Formula Weight 291.2
MDL Number MFCD16294544
Synonym 2-(Methyl[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]amino)ethanol