Please login to your online account to display your discounted pricing

4-(2-Hydroxyethyl)benzonitrile, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  TL00139CB

 View more versions of this product

Catalog No. TL00139CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 147.18
Quantity 250mg
Assay 97%
Molecular Formula C9H9NO
Linear Formula CNC6H4(CH2)2OH
Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile
CAS 69395-13-7
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NO
Linear Formula CNC6H4(CH2)2OH
Formula Weight 147.18