Learn More
4-(2-Hydroxyethyl)benzonitrile, 98%, Thermo Scientific Chemicals
Organic intermediate
Supplier: Thermo Scientific Chemicals 41283MD
Description
4-(2-Hydroxyethyl)benzonitrile is used to produce 4-(2-bromo-ethyl)-benzonitrile. It is used as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(2-Hydroxyethyl)benzonitrile | |
116°C to 118°C (0.5 mmHg) | |
MFCD00052461 | |
UN3439 | |
RBSJBNYPTGMZIH-UHFFFAOYSA-N | |
4-(2-hydroxyethyl)benzonitrile | |
2800815 | |
98% |
69395-13-7 | |
C9H9NO | |
250 mg | |
4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6 | |
C1=CC(=CC=C1CCO)C#N | |
147.177 | |
147.18 | |
Chunks and/or Powder |
Chemical Identifiers
69395-13-7 | |
147.177 | |
RBSJBNYPTGMZIH-UHFFFAOYSA-N | |
2800815 | |
C1=CC(=CC=C1CCO)C#N |
C9H9NO | |
MFCD00052461 | |
4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6 | |
4-(2-hydroxyethyl)benzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only