missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[4-(2-Furyl)phenyl]methanol, ≥97%, Maybridge

Chemical Identifiers

CAS 17920-85-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H10O2
Formula Weight 174.2
MDL Number MFCD06203036
Chemical Name or Material [4-(2-Furyl)phenyl]methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC34409EA Fisher Scientific Company
CC34409EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34409DA SDS Fisher Scientific Company
CC34409DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34409CB SDS Fisher Scientific Company
CC34409CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17920-85-3
100.0
C11H10O2
MFCD06203036
97%
96.5
97%
174.2
[4-(2-Furyl)phenyl]methanol

Specifications

17920-85-3
100.0
C11H10O2
MFCD06203036
[4-(2-Furyl)phenyl]methanol
96.5
97%
174.2
10g
97%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.