Please login to your online account to display your discounted pricing

4-(2-furyl)benzaldehyde, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC34404CB

 View more versions of this product

Catalog No. CC34404CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.18
Quantity 250mg
Assay 97%
Molecular Formula C11H8O2
Chemical Name or Material 4-(2-furyl)benzaldehyde
CAS 60456-77-1

Specifications

Chemical Name or Material 4-(2-furyl)benzaldehyde
Assay 97%
Molecular Formula C11H8O2
Formula Weight 172.18