Please login to your online account to display your discounted pricing

4-(2-Furyl)aniline, ≥97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC34414DA

 View more versions of this product

Catalog No. CC34414DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 159.19
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9NO
MDL Number MFCD04039072
CAS 59147-02-3
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Chemical Name or Material 4-(2-Furyl)aniline
Packaging Amber glass bottle
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(2-Furyl)aniline
CAS 59147-02-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9NO
Formula Weight 159.19
MDL Number MFCD04039072
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.

SDS & Safety Documents