Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Formylphenoxy)benzenesulfonamide

Chemical Identifiers

Chemical Name or Material 4-(2-Formylphenoxy)benzenesulfonamide
CAS 902837-00-7
Molecular Formula C13H11NO4S
Formula Weight 277.3
MDL Number MFCD08061110
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5100503

 
alfa aesar™
H5100503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-(2-Formylphenoxy)benzenesulfonamide
Quantity 1g
CAS 902837-00-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H11NO4S
Formula Weight 277.3
MDL Number MFCD08061110

Applications
It is employed as intermediate for pharmaceutical and agrochemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.