Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Fluorophenyl)-3-thiosemicarbazide, 96%

Chemical Identifiers

Chemical Name or Material 4-(2-Fluorophenyl)-3-thiosemicarbazide
CAS 38985-72-7
Molecular Formula C7H8FN3S
Formula Weight 185.22
MDL Number MFCD00041269
UN Number UN2811
Beilstein 2212143
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1292006

  SDS
alfa aesar™
L1292006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.22
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 1 Danger
Chemical Name or Material 4-(2-Fluorophenyl)-3-thiosemicarbazide
CAS 38985-72-7
Quantity 5g
Assay 96%
Molecular Formula C7H8FN3S
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
MDL Number MFCD00041269
Beilstein 2212143
UN Number UN2811
Melting Point ≈156°C
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.