missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Fluorobenzyl)piperidine hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H63849MD

 View more versions of this product

Catalog No. AAH63849MD



Specifications

Specifications

4-(2-Fluorobenzyl)piperidine hydrochloride
250mg
C12H17ClFN
4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl
C1CNCCC1CC2=CC(=CC=C2)F.Cl
229.723
229.72
97%
White
193357-21-0
MFCD09787475
XZRQMBCQGGXDKQ-UHFFFAOYSA-N
4-[(3-fluorophenyl)methyl]piperidine;hydrochloride
17039493
Solid

Chemical Identifiers

193357-21-0
229.723
XZRQMBCQGGXDKQ-UHFFFAOYSA-N
17039493
C1CNCCC1CC2=CC(=CC=C2)F.Cl
C12H17ClFN
MFCD09787475
4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl
4-[(3-fluorophenyl)methyl]piperidine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents