missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Fluorobenzyl)piperidine hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H6384906

 View more versions of this product

Catalog No. AAH6384906



Specifications

Specifications

4-(2-Fluorobenzyl)piperidine hydrochloride
5g
C12H17ClFN
4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl, 4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride
C1CNCCC1CC2=CC(=CC=C2)F.Cl
229.723
229.72
97%
White
193357-21-0
MFCD09787475
XZRQMBCQGGXDKQ-UHFFFAOYSA-N
4-[(3-fluorophenyl)methyl]piperidine;hydrochloride
17039493
Solid

Chemical Identifiers

193357-21-0
229.723
XZRQMBCQGGXDKQ-UHFFFAOYSA-N
17039493
C1CNCCC1CC2=CC(=CC=C2)F.Cl
C12H17ClFN
MFCD09787475
4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl, 4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride
4-[(3-fluorophenyl)methyl]piperidine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents