missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(2-Fluorobenzyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6384903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(2-Fluorobenzyl)piperidine hydrochloride | |
White | |
MFCD08669673 | |
4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl | |
Cl.FC1=CC=CC(CC2CCNCC2)=C1 | |
229.72 | |
229.72 | |
Solid |
193357-21-0 | |
C12H17ClFN | |
1 g | |
XZRQMBCQGGXDKQ-UHFFFAOYSA-N | |
4-[(3-fluorophenyl)methyl]piperidine hydrochloride | |
17039493 | |
97% |
Chemical Identifiers
193357-21-0 | |
229.72 | |
XZRQMBCQGGXDKQ-UHFFFAOYSA-N | |
17039493 |
C12H17ClFN | |
MFCD08669673 | |
4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl | |
Cl.FC1=CC=CC(CC2CCNCC2)=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only