missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Fluoro-6-nitrophenyl)morpholine, 97%

Manufacturer:  Alfa Aesar™ H6327206

 View more versions of this product

Catalog No. AAH6327206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-(2-Fluoro-6-nitrophenyl)morpholine
1233955-40-2
MFCD13689260
RVPOFXBMBJKJJK-UHFFFAOYSA-N
4-(2-fluoro-6-nitrophenyl)morpholine
53255510
Solid
5g
C10H11FN2O3
4-2-fluoro-6-nitrophenyl morpholine
C1COCCN1C2=C(C=CC=C2F)[N+](=O)[O-]
226.207
226.2
97%

Chemical Identifiers

1233955-40-2
226.207
RVPOFXBMBJKJJK-UHFFFAOYSA-N
53255510
C1COCCN1C2=C(C=CC=C2F)[N+](=O)[O-]
C10H11FN2O3
MFCD13689260
4-2-fluoro-6-nitrophenyl morpholine
4-(2-fluoro-6-nitrophenyl)morpholine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents