missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-{[2-(Dimethylamino)ethyl]carbamoyl}phenylboronic acid pinacol ester, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444940010

 View more versions of this product

Catalog No. AC444940010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-{[2-(Dimethylamino)ethyl]carbamoyl}phenylboronic acid pinacol ester
107°C to 110°C
832114-11-1
C17H27BN2O3
KTDBEJMIGCLCSU-UHFFFAOYSA-N
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
2758503
95%
Glass Bottle
1g
94% min. (GC)
4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester, amtb351, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
318.23
318.23

Chemical Identifiers

832114-11-1
318.23
4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester, amtb351, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
C17H27BN2O3
KTDBEJMIGCLCSU-UHFFFAOYSA-N
2758503
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents