missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-{[2-(Dimethylamino)ethyl]carbamoyl}phenylboronic acid pinacol ester, 95%, ACROS Organics™

Chemical Identifiers

CAS 832114-11-1
Molecular Formula C17H27BN2O3
Molecular Weight (g/mol) 318.23
InChI Key KTDBEJMIGCLCSU-UHFFFAOYSA-N
Synonym 4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester
PubChem CID 2758503
IUPAC Name N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444940010 SDS View Documents Thermo Fisher Scientific
AC444940010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444940050 SDS View Documents Thermo Fisher Scientific
AC444940050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

832114-11-1
318.23
4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
C17H27BN2O3
KTDBEJMIGCLCSU-UHFFFAOYSA-N
2758503
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C

Specifications

Glass Bottle
1g
94% min. (GC)
4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
318.23
318.23
4-{[2-(Dimethylamino)ethyl]carbamoyl}phenylboronic acid pinacol ester
107°C to 110°C
832114-11-1
C17H27BN2O3
KTDBEJMIGCLCSU-UHFFFAOYSA-N
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
2758503
95%
Safety and Handling

Safety and Handling

GHS Signal Word: Danger