missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

Chemical Identifiers

Molecular Formula C18H22BNO4
Formula Weight 327.18
MDL Number MFCD11113044
Synonym Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate
Chemical Name or Material 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5113003 SDS Alfa Aesar™
H5113003
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5113006 SDS Alfa Aesar™
H5113006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5113014 SDS Alfa Aesar™
H5113014
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C18H22BNO4
MFCD11113044
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester
327.18
Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate

Specifications

1g
327.18
Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate
C18H22BNO4
MFCD11113044
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning