Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chlorophenyl)tetrahydropyran-4-carboxaldehyde, 98%

$221.26 - $221.26

Chemical Identifiers

Chemical Name or Material 4-(2-Chlorophenyl)tetrahydropyran-4-carboxaldehyde
CAS 902836-47-9
Molecular Formula C12H13ClO2
Formula Weight 224.69g/mol
MDL Number MFCD08061003
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5081103

 
alfa aesar™
H50811-03
Each for $221.26
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-(2-Chlorophenyl)tetrahydropyran-4-carboxaldehyde
CAS 902836-47-9
Assay 98%
Molecular Formula C12H13ClO2
Formula Weight 224.69g/mol
MDL Number MFCD08061003
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.