Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chlorophenoxy)piperidine hydrochloride

$90.81 - $327.86

Chemical Identifiers

Chemical Name or Material 4-(2-Chlorophenoxy)piperidine hydrochloride
CAS 849107-20-6
Molecular Formula C11H14ClNO·HCl
Formula Weight 248.15
MDL Number MFCD02684107
Synonym 4-(2-Chlorophenoxy)piperidinium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51029MD

 
alfa aesar™
H51029MD
250mg Each for $90.81

AAH5102903

 
alfa aesar™
H5102903
1g Each for $327.86
Description & Specifications

Specifications

Formula Weight 248.15
Molecular Formula C11H14ClNO·HCl
MDL Number MFCD02684107
Synonym 4-(2-Chlorophenoxy)piperidinium chloride
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-(2-Chlorophenoxy)piperidine hydrochloride
CAS 849107-20-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.