Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chloroethyl)morpholine hydrochloride, 99%

Chemical Identifiers

Chemical Name or Material 4-(2-Chloroethyl)morpholine hydrochloride
CAS 3647-69-6
Molecular Formula C6H12ClNO·HCl
Formula Weight 186.08
MDL Number MFCD00012797
UN Number UN2923
Beilstein 3684083
Synonym 2-(4-Morpholinyl)ethyl chloride hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0577122

  SDS
alfa aesar™
L0577122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0577136

  SDS
alfa aesar™
L0577136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.08
Chemical Name or Material 4-(2-Chloroethyl)morpholine hydrochloride
CAS 3647-69-6
Health Hazard 2 GHS H Statement
H301-H314-H318-H312-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
TSCA TSCA
Assay 99%
Molecular Formula C6H12ClNO·HCl
MDL Number MFCD00012797
UN Number UN2923
Beilstein 3684083
Synonym 2-(4-Morpholinyl)ethyl chloride hydrochloride
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
RTECS Number QE0260000
Melting Point 184° to 186°C
Quantity 100g

Applications
4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Hygroscopic. Do not store together with acids. Store under dry inert gas. Protect from humidity and water. Incompatible materials are acids, moisture.

Safety and Handling

Danger

GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H314-H318-H312-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.