Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(2-Chloroethyl)acetophenone, tech. 90%

$144.80 - $673.53

Chemical Identifiers

Chemical Name or Material 4'-(2-Chloroethyl)acetophenone
CAS 69614-95-5
Purity Grade Technical Grade
Molecular Formula C10H11ClO
Formula Weight 182.65
MDL Number MFCD00016350
Beilstein 2081019
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1380006

  SDS
alfa aesar™
L1380006
5g Each for $144.80

AAL1380014

  SDS
alfa aesar™
L1380014
25g Each for $673.53
Description & Specifications

Specifications

Formula Weight 182.65
CAS 69614-95-5
Assay 90%
Chemical Name or Material 4'-(2-Chloroethyl)acetophenone
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C10H11ClO
Beilstein 2081019
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Purity Grade Technical Grade
Quantity 5g
Boiling Point 112°C (0.1mmHg)
MDL Number MFCD00016350
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.