missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(2-Chloroethyl)acetophenone, tech. 90%

Manufacturer:  Alfa Aesar™ L1380006

 View more versions of this product

Catalog No. AAL1380006



Specifications

Specifications

4'-(2-Chloroethyl)acetophenone
5g
C10H11ClO
2081019
RGVUACHOYCYNMB-UHFFFAOYSA-N
1-[4-(2-chloroethyl)phenyl]ethanone
144359
90%
112°C (0.1mmHg)
69614-95-5
MFCD00016350
4'-beta-chloroethyl acetophenone, 4-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 4'-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl-1-ethanone, 4'-.beta.-chloroethyl acetophenone, p-beta-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 1-4-2-chloroethyl phenyl ethan-1-one, 1-acetyl-4-2-chloroethyl benzene
CC(=O)C1=CC=C(C=C1)CCCl
182.647
182.65
Technical

Chemical Identifiers

69614-95-5
182.647
RGVUACHOYCYNMB-UHFFFAOYSA-N
144359
CC(=O)C1=CC=C(C=C1)CCCl
C10H11ClO
MFCD00016350
4'-beta-chloroethyl acetophenone, 4-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 4'-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl-1-ethanone, 4'-.beta.-chloroethyl acetophenone, p-beta-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 1-4-2-chloroethyl phenyl ethan-1-one, 1-acetyl-4-2-chloroethyl benzene
1-[4-(2-chloroethyl)phenyl]ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents