missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chloroacetyl)-2H-1,4-benzothiazin-3(4H)-one, 97%

$43.03 - $135.85

Chemical Identifiers

CAS 68321-41-5
Molecular Formula C10H8ClNO2S
Molecular Weight (g/mol) 241.689
MDL Number MFCD23379849
InChI Key CYIZUSMRAHFMOI-UHFFFAOYSA-N
Synonym 4-2-chloroacetyl-2h-1,4-benzothiazin-3 4h-one, 4-2-chloroacetyl-2h-1,4-benzothiazin-3-one
PubChem CID 12445921
IUPAC Name 4-(2-chloroacetyl)-1,4-benzothiazin-3-one
SMILES C1C(=O)N(C2=CC=CC=C2S1)C(=O)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61022MD View Documents Alfa Aesar™
H61022MD
250mg Each for $43.03
 
AAH6102203 View Documents Alfa Aesar™
H6102203
1g Each for $135.85
 
Specifications

Chemical Identifiers

68321-41-5
241.689
CYIZUSMRAHFMOI-UHFFFAOYSA-N
12445921
C1C(=O)N(C2=CC=CC=C2S1)C(=O)CCl
C10H8ClNO2S
MFCD23379849
4-2-chloroacetyl-2h-1,4-benzothiazin-3 4h-one, 4-2-chloroacetyl-2h-1,4-benzothiazin-3-one
4-(2-chloroacetyl)-1,4-benzothiazin-3-one

Specifications

250mg
C10H8ClNO2S
4-2-chloroacetyl-2h-1,4-benzothiazin-3 4h-one, 4-2-chloroacetyl-2h-1,4-benzothiazin-3-one
C1C(=O)N(C2=CC=CC=C2S1)C(=O)CCl
241.689
241.69
4-(2-Chloroacetyl)-2H-1,4-benzothiazin-3(4H)-one
68321-41-5
MFCD23379849
CYIZUSMRAHFMOI-UHFFFAOYSA-N
4-(2-chloroacetyl)-1,4-benzothiazin-3-one
12445921
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning