Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chloro-6-nitrophenyl)morpholine, 97%

Chemical Identifiers

Chemical Name or Material 4-(2-Chloro-6-nitrophenyl)morpholine
CAS 65152-10-5
Formula Weight 242.66
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6374403

 
alfa aesar™
H6374403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6374406

 
alfa aesar™
H6374406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63744MD

 
alfa aesar™
H63744MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.66
Chemical Name or Material 4-(2-Chloro-6-nitrophenyl)morpholine
Physical Form Solid
Assay 97%
CAS 65152-10-5
Quantity 1g