Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chloro-5-methylphenoxy)piperidine, 99%

$104.53 - $363.27

Chemical Identifiers

Chemical Name or Material 4-(2-Chloro-5-methylphenoxy)piperidine
CAS 254883-43-7
Molecular Formula C12H16ClNO
Formula Weight 225.72
MDL Number MFCD06246346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5077803

 
alfa aesar™
H5077803
1g Each for $363.27

AAH50778MD

 
alfa aesar™
H50778MD
250mg Each for $104.53
Description & Specifications

Specifications

Formula Weight 225.72
Assay 99%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-(2-Chloro-5-methylphenoxy)piperidine
Molecular Formula C12H16ClNO
MDL Number MFCD06246346
CAS 254883-43-7
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.