Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Chloro-4-nitrophenyl)morpholine, 97%

Chemical Identifiers

Chemical Name or Material 4-(2-Chloro-4-nitrophenyl)morpholine
CAS 55435-71-7
Formula Weight 242.66
MDL Number MFCD00052898
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6306303

 
alfa aesar™
H6306303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6306306

 
alfa aesar™
H6306306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63063MD

 
alfa aesar™
H63063MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.66
Chemical Name or Material 4-(2-Chloro-4-nitrophenyl)morpholine
TSCA TSCA
Physical Form Solid
Assay 97%
MDL Number MFCD00052898
CAS 55435-71-7
Quantity 1g