Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Carboxyvinyl)benzeneboronic acid, 95%

$82.50 - $218.58

Chemical Identifiers

Chemical Name or Material 4-(2-Carboxyvinyl)benzeneboronic acid
CAS 151169-68-5
Molecular Formula C9H9BO4
Formula Weight 191.98
MDL Number MFCD01075704
Beilstein 9196727
Synonym 4-Boronocinnamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1558603

 
alfa aesar™
L1558603
1g Each for $218.58

AAL15586MD

 
alfa aesar™
L15586MD
250mg Each for $82.50
Description & Specifications

Specifications

Formula Weight 191.98
Chemical Name or Material 4-(2-Carboxyvinyl)benzeneboronic acid
Assay 95%
Molecular Formula C9H9BO4
MDL Number MFCD01075704
Health Hazard 1 Warning
Beilstein 9196727
Synonym 4-Boronocinnamic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈190° to 196°C
Quantity 1g
CAS 151169-68-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.