missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5309503

 View more versions of this product

Catalog No. AAH5309503



Specifications

Specifications

4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid
1g
C8H10BN3O3S
2-4-boronobenzoyl hydrazinecarbothioamide, 4-2-carbamothioylhydrazinecarbonyl phenylboronic acid, 4-2-carbamothioylhydrazino carbonyl benzeneboronic acid, 4-carbamothioylaminocarbamoyl phenylboronic acid, 4-carbamothioylamino carbamoyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenylboronic
B(C1=CC=C(C=C1)C(=O)NNC(=S)N)(O)O
239.056
239.06
183°C to 184°C
957060-76-3
MFCD09027244
NVXNXTLWXKMYHB-UHFFFAOYSA-N
[4-[(carbamothioylamino)carbamoyl]phenyl]boronic acid
44119176
98%

Chemical Identifiers

957060-76-3
239.056
NVXNXTLWXKMYHB-UHFFFAOYSA-N
44119176
B(C1=CC=C(C=C1)C(=O)NNC(=S)N)(O)O
C8H10BN3O3S
MFCD09027244
2-4-boronobenzoyl hydrazinecarbothioamide, 4-2-carbamothioylhydrazinecarbonyl phenylboronic acid, 4-2-carbamothioylhydrazino carbonyl benzeneboronic acid, 4-carbamothioylaminocarbamoyl phenylboronic acid, 4-carbamothioylamino carbamoyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenylboronic
[4-[(carbamothioylamino)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents