missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Bromophenyl)-2-methyl-1,3-thiazole, 97%, Maybridge

Chemical Identifiers

CAS 941717-01-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H8BrNS
Formula Weight 254.15
MDL Number MFCD09879981
Chemical Name or Material 4-(2-Bromophenyl)-2-methyl-1, 3-thiazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC68410DA SDS Fisher Scientific Company
CC68410DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC68410DE SDS Fisher Scientific Company
CC68410DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

941717-01-7
100.0
C10H8BrNS
MFCD09879981
97%
96.5
97%
254.15
4-(2-Bromophenyl)-2-methyl-1, 3-thiazole

Specifications

941717-01-7
100.0
1g
C10H8BrNS
MFCD09879981
97%
96.5
Amber Glass Bottle
97%
254.15
4-(2-Bromophenyl)-2-methyl-1, 3-thiazole

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning