missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
$98.51 - $320.38
Chemical Identifiers
CAS | 913836-27-8 |
---|---|
Molecular Formula | C14H20BBrO3 |
Molecular Weight (g/mol) | 327.03 |
MDL Number | MFCD09027285 |
InChI Key | NTTOMYNPPZBRBG-UHFFFAOYSA-N |
Synonym | 2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-2-bromoethoxy phenylboronic acid, pinacol ester, 4-2-bromoethoxy benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl |
PubChem CID | 44119155 |
IUPAC Name | 2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=C(OCCBr)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH53246MD
|
Thermo Scientific Chemicals
H53246MD |
250 mg |
Each for $98.51
|
|
|||||
AAH5324603
|
Thermo Scientific Chemicals
H5324603 |
1 g |
Each for $320.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
913836-27-8 | |
327.03 | |
NTTOMYNPPZBRBG-UHFFFAOYSA-N | |
44119155 | |
CC1(C)OB(OC1(C)C)C1=CC=C(OCCBr)C=C1 |
C14H20BBrO3 | |
MFCD09027285 | |
2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-2-bromoethoxy phenylboronic acid, pinacol ester, 4-2-bromoethoxy benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl | |
2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
913836-27-8 | |
C14H20BBrO3 | |
250 mg | |
NTTOMYNPPZBRBG-UHFFFAOYSA-N | |
2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
44119155 | |
98% |
90°C to 92°C | |
MFCD09027285 | |
2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-2-bromoethoxy phenylboronic acid, pinacol ester, 4-2-bromoethoxy benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl | |
CC1(C)OB(OC1(C)C)C1=CC=C(OCCBr)C=C1 | |
327.03 | |
327.03 | |
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only