missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Bromoethoxy)benzeneboronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H53246MD

 View more versions of this product

Catalog No. AAH53246MD



Specifications

Specifications

4-(2-Bromoethoxy)benzeneboronic acid pinacol ester
250mg
C14H20BBrO3
2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-2-bromoethoxy phenylboronic acid, pinacol ester, 4-2-bromoethoxy benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)OCCBr
327.025
327.03
90°C to 92°C
913836-27-8
MFCD09027285
NTTOMYNPPZBRBG-UHFFFAOYSA-N
2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
44119155
98%

Chemical Identifiers

913836-27-8
327.025
NTTOMYNPPZBRBG-UHFFFAOYSA-N
44119155
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)OCCBr
C14H20BBrO3
MFCD09027285
2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-2-bromoethoxy phenylboronic acid, pinacol ester, 4-2-bromoethoxy benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-2-bromoethoxy phenyl-4,4,5,5-tetramethyl
2-[4-(2-bromoethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SDS Documents