Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Bromo-6-fluorobenzyl)morpholine, 96%

Chemical Identifiers

Chemical Name or Material 4-(2-Bromo-6-fluorobenzyl)morpholine
Formula Weight 274.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6222903

 
alfa aesar™
H6222903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6222906

 
alfa aesar™
H6222906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62229MD

 
alfa aesar™
H62229MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.13
Quantity 1g
Physical Form Solid
Assay 96%
Chemical Name or Material 4-(2-Bromo-6-fluorobenzyl)morpholine