Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Bromo-5-fluorobenzyl)morpholine, 96%

Chemical Identifiers

Chemical Name or Material 4-(2-Bromo-5-fluorobenzyl)morpholine
Formula Weight 274.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6257403

 
alfa aesar™
H6257403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62574MD

 
alfa aesar™
H62574MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.13
Physical Form Solid
Assay 96%
Quantity 1g
Chemical Name or Material 4-(2-Bromo-5-fluorobenzyl)morpholine