Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Bromo-4-fluorobenzyl)morpholine, 96%

Chemical Identifiers

Chemical Name or Material 4-(2-Bromo-4-fluorobenzyl)morpholine
CAS 1086600-40-9
Formula Weight 274.13
MDL Number MFCD14647294
Beilstein 18768963
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6235903

 
alfa aesar™
H6235903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6235906

 
alfa aesar™
H6235906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62359MD

 
alfa aesar™
H62359MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.13
Assay 96%
MDL Number MFCD14647294
CAS 1086600-40-9
Physical Form Solid
Quantity 1g
Beilstein 18768963
Chemical Name or Material 4-(2-Bromo-4-fluorobenzyl)morpholine