Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[2-(Boc-amino)ethyl]aniline, 97%

Chemical Identifiers

Chemical Name or Material 4-[2-(Boc-amino)ethyl]aniline
CAS 94838-59-2
Formula Weight 236.31
MDL Number MFCD05663961
UN Number UN2811
Synonym tert-Butyl 4-aminophenethylcarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6332403

  SDS
alfa aesar™
H6332403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6332406

  SDS
alfa aesar™
H6332406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63324MD

  SDS
alfa aesar™
H63324MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.31
Chemical Name or Material 4-[2-(Boc-amino)ethyl]aniline
Physical Form Solid
Assay 97%
MDL Number MFCD05663961
Quantity 1g
CAS 94838-59-2
UN Number UN2811
Color Brown
Synonym tert-Butyl 4-aminophenethylcarbamate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate -20°C. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C