Learn More
4-[2-(Boc-amino)ethyl]aniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6332406
Description
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.
Solubility
Slightly soluble in water.
Notes
Refrigerate -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications
4-[2-(Boc-amino)ethyl]aniline | |
Brown | |
MFCD05663961 | |
tert-butyl 4-aminophenethylcarbamate, 4-2-boc-amino ethyl aniline, 2-4-amino-phenyl-ethyl-carbamic acid tert-butyl ester, tert-butyl n-2-4-aminophenyl ethyl carbamate, 2-4-aminophenyl ethyl carbamic acid tert-butyl ester, tert-butyl 4-aminophenethyl carbamate, tert-butyl-2-4-aminophenyl ethylcarbamate, 2-methyl-2-propanyl 2-4-aminophenyl ethyl carbamate, carbamic acid, n-2-4-aminophenyl ethyl-, 1,1-dimethylethyl ester, acmc-209ru6 | |
HOPALBZGTWDOTL-UHFFFAOYSA-N | |
tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate | |
382207 | |
97% | |
Solid |
94838-59-2 | |
C13H20N2O2 | |
UN2811 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)NCCC1=CC=C(C=C1)N | |
236.315 | |
236.31 | |
5 g |
Chemical Identifiers
94838-59-2 | |
236.315 | |
HOPALBZGTWDOTL-UHFFFAOYSA-N | |
382207 | |
CC(C)(C)OC(=O)NCCC1=CC=C(C=C1)N |
C13H20N2O2 | |
MFCD05663961 | |
tert-butyl 4-aminophenethylcarbamate, 4-2-boc-amino ethyl aniline, 2-4-amino-phenyl-ethyl-carbamic acid tert-butyl ester, tert-butyl n-2-4-aminophenyl ethyl carbamate, 2-4-aminophenyl ethyl carbamic acid tert-butyl ester, tert-butyl 4-aminophenethyl carbamate, tert-butyl-2-4-aminophenyl ethylcarbamate, 2-methyl-2-propanyl 2-4-aminophenyl ethyl carbamate, carbamic acid, n-2-4-aminophenyl ethyl-, 1,1-dimethylethyl ester, acmc-209ru6 | |
tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only