missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[2-(Boc-amino)ethyl]aniline, 97%

Manufacturer:  Alfa Aesar™ H63324MD

 View more versions of this product

Catalog No. AAH63324MD



Description

Description

Applications
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

4-[2-(Boc-amino)ethyl]aniline
250mg
C13H20N2O2
UN2811
HOPALBZGTWDOTL-UHFFFAOYSA-N
tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate
382207
Solid
Brown
94838-59-2
MFCD05663961
tert-butyl 4-aminophenethylcarbamate, 4-2-boc-amino ethyl aniline, 2-4-amino-phenyl-ethyl-carbamic acid tert-butyl ester, tert-butyl n-2-4-aminophenyl ethyl carbamate, 2-4-aminophenyl ethyl carbamic acid tert-butyl ester, tert-butyl 4-aminophenethyl carbamate, tert-butyl-2-4-aminophenyl ethylcarbamate, 2-methyl-2-propanyl 2-4-aminophenyl ethyl carbamate, carbamic acid, n-2-4-aminophenyl ethyl-, 1,1-dimethylethyl ester, acmc-209ru6
CC(C)(C)OC(=O)NCCC1=CC=C(C=C1)N
236.315
236.31
97%

Chemical Identifiers

94838-59-2
236.315
HOPALBZGTWDOTL-UHFFFAOYSA-N
382207
CC(C)(C)OC(=O)NCCC1=CC=C(C=C1)N
C13H20N2O2
MFCD05663961
tert-butyl 4-aminophenethylcarbamate, 4-2-boc-amino ethyl aniline, 2-4-amino-phenyl-ethyl-carbamic acid tert-butyl ester, tert-butyl n-2-4-aminophenyl ethyl carbamate, 2-4-aminophenyl ethyl carbamic acid tert-butyl ester, tert-butyl 4-aminophenethyl carbamate, tert-butyl-2-4-aminophenyl ethylcarbamate, 2-methyl-2-propanyl 2-4-aminophenyl ethyl carbamate, carbamic acid, n-2-4-aminophenyl ethyl-, 1,1-dimethylethyl ester, acmc-209ru6
tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents