Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Aminoethyl)pyridine, 97%

Chemical Identifiers

Chemical Name or Material 4-(2-Aminoethyl)pyridine
CAS 13258-63-4
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00038045
UN Number UN2735
Synonym 4-Pyridineethylamine; 2-(4-Pyridyl)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2754503

  SDS
alfa aesar™
H2754503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2754506

  SDS
alfa aesar™
H2754506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material 4-(2-Aminoethyl)pyridine
TSCA TSCA
Boiling Point 70° to 71°C (1mmHg)
Assay 97%
Quantity 1g
Flash Point >110°C (230°F)
Odor Amine-like
Molecular Formula C7H10N2
MDL Number MFCD00038045
CAS 13258-63-4
UN Number UN2735
Synonym 4-Pyridineethylamine; 2-(4-Pyridyl)ethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.