Learn More
4-(2-Aminoethyl)morpholine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 400751000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(2-Aminoethyl)morpholine | |
98.5 | |
24°C | |
0.9900g/mL | |
175°C | |
98% min. (GC) | |
C6H14N2O | |
MFCD00006182 | |
0.99 | |
Solubility in water: soluble | |
C1COCCN1CCN | |
130.19 | |
130.19 | |
Liquid After Melting |
2038-03-1 | |
100.0 | |
Colorless to Yellow | |
205°C | |
Authentic | |
Glass bottle | |
1.4745 to 1.4765 | |
100 g | |
4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine | |
RWIVICVCHVMHMU-UHFFFAOYSA-N | |
2-morpholin-4-ylethanamine | |
408285 | |
98+% |
Chemical Identifiers
2038-03-1 | |
130.19 | |
RWIVICVCHVMHMU-UHFFFAOYSA-N |
C6H14N2O | |
MFCD00006182 |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 218-011-6
RUO – Research Use Only