Please login to your online account to display your discounted pricing

4-(2-Aminoethyl)benzenesulfonyl Fluoride HCl (White Powder), Fisher BioReagents


Quantity 10mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP264410

 View more versions of this product

Catalog No. BP264410

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 239.7
Melting Point 175°C
Physical Form Powder/Solid
Quantity 10mg
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Color White
Molecular Formula C8H10FNO2S · HCl
CAS 30827-99-7
Health Hazard 3 Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:
Recommended Storage RT, protect from light.
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
Purity 97% min.
Assay Percent Range ≥97 %
Synonym AEBSF
Infrared Spectrum Pass Test
Identification Pass Test
Packaging Amber Glass
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:DANGER!

Specifications

Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
CAS 30827-99-7
Physical Form Powder/Solid
Assay Percent Range ≥97 %
Molecular Formula C8H10FNO2S · HCl
Formula Weight 239.7
Synonym AEBSF

  • AEBSF is  water-soluble, irreversible serine protease inhibitor
  • Relative to PMSF, AEBSF is more soluble in water, and provides lower toxicity and higher inhibitory activity
  • Sulfonyl fluoride that reacts with the hydroxy group of the active site serine residue
  • Typical concentrations for use are 0.1 - 1.0 mM

Accessories
 

Signal Word:DANGER!

Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:

RT, protect from light.

SDS & Safety Documents